Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711411.3 | XP_002711457.1 | 5 - 6 | 1962 | 61 | |
| XM_008257081.2 | XP_008255303.1 | 5 - 6 | 1962 | 61 | ||
More Information
Set Membership: |
|

FOXRED2
Genomic Map