Assay Details
| Gene Symbol: NMI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258576.2 | XP_008256798.1 | 3 - 4 | 318 | 75 | |
| XM_017342844.1 | XP_017198333.1 | 2 - 3 | 315 | 75 | ||
| XM_017342845.1 | XP_017198334.1 | 3 - 4 | 322 | 75 | ||
More Information
Set Membership: |
|

NMI
Genomic Map