Assay Details
| Gene Symbol: ZFYVE27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718607.3 | XP_002718653.1 | 2 - 3 | 397 | 61 | |
| XM_002718608.3 | XP_002718654.1 | 2 - 3 | 396 | 61 | ||
| XM_008270306.2 | XP_008268528.1 | 2 - 3 | 399 | 61 | ||
| XM_008270307.2 | XP_008268529.1 | 2 - 3 | 443 | 61 | ||
| XM_008270308.2 | XP_008268530.1 | 2 - 3 | 397 | 61 | ||
| XM_017348474.1 | XP_017203963.1 | 2 - 3 | 217 | 61 | ||
More Information
Set Membership: |
|

ZFYVE27
Genomic Map