Assay Details
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710540.3 | XP_002710586.1 | 25 - 26 | 3331 | 59 | |
| XM_008255901.2 | XP_008254123.1 | 26 - 27 | 3411 | 59 | ||
More Information
Set Membership: |
|

WASHC5
Genomic Map