Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711334.3 | XP_002711380.2 | 6 - 7 | 1000 | 74 | |
| XM_008256790.2 | XP_008255012.1 | 7 - 8 | 1092 | 74 | ||
More Information
Set Membership: |
|

TMEM19
Genomic Map