Assay Details
| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345583.1 | XP_017201072.1 | 5 - 6 | 528 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGBL4| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345583.1 | XP_017201072.1 | 5 - 6 | 528 | 57 | |
Set Membership: |
|