Assay Details
| Gene Symbol: ZNF143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266017.2 | XP_008264239.2 | 6 - 7 | 659 | 65 | |
| XM_008266024.2 | XP_008264246.2 | 6 - 7 | 656 | 65 | ||
| XM_008266029.2 | XP_008264251.1 | 6 - 7 | 664 | 65 | ||
| XM_008266037.2 | XP_008264259.1 | 6 - 7 | 673 | 65 | ||
| XM_017345320.1 | XP_017200809.1 | 7 - 8 | 832 | 65 | ||
More Information
Set Membership: |
|

ZNF143
Genomic Map