Assay Details
| Gene Symbol: TCP11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711428.3 | XP_002711474.1 | 4 - 5 | 582 | 74 | |
| XM_017342128.1 | XP_017197617.1 | 3 - 4 | 407 | 74 | ||
| XM_017342130.1 | XP_017197619.1 | 3 - 4 | 407 | 74 | ||
More Information
Set Membership: |
|

TCP11L2
Genomic Map