Assay Details
Gene Symbol: DEDD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715205.3 | XP_002715251.1 | 4 - 5 | 1887 | 55 | |
XM_017345740.1 | XP_017201229.1 | 5 - 6 | 1317 | 55 | ||
XM_017345741.1 | XP_017201230.1 | 5 - 6 | 742 | 55 | ||
XM_017345742.1 | XP_017201231.1 | 4 - 5 | 1284 | 55 | ||
XM_017345743.1 | XP_017201232.1 | 5 - 6 | 1130 | 55 | ||
XM_017345744.1 | XP_017201233.1 | 4 - 5 | 731 | 55 | ||
XM_017345745.1 | XP_017201234.1 | 4 - 5 | 821 | 55 | ||
XM_017345746.1 | XP_017201235.1 | 5 - 6 | 1097 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
