Assay Details
| Gene Symbol: CNNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718589.3 | XP_002718635.1 | 6 - 7 | 2482 | 61 | |
| XM_002718590.3 | XP_002718636.1 | 7 - 8 | 2415 | 61 | ||
| XM_008270432.2 | XP_008268654.1 | 8 - 9 | 2667 | 61 | ||
| XM_008270433.2 | XP_008268655.1 | 7 - 8 | 2601 | 61 | ||
More Information
Set Membership: |
|

CNNM2
Genomic Map