Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721876.3 | XP_002721922.1 | 666 | 73 | ||
| XM_008248907.2 | XP_008247129.1 | 670 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM4| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721876.3 | XP_002721922.1 | 666 | 73 | ||
| XM_008248907.2 | XP_008247129.1 | 670 | 73 | |||
Set Membership: |
|