Assay Details
| Gene Symbol: EEF1AKMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720946.3 | XP_002720992.1 | 533 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EEF1AKMT3| Gene Symbol: EEF1AKMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720946.3 | XP_002720992.1 | 533 | 59 | ||
Set Membership: |
|