Assay Details
| Gene Symbol: LOC100340625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002707963.3 | XP_002708009.2 | 1 - 2 | 195 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100340625| Gene Symbol: LOC100340625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002707963.3 | XP_002708009.2 | 1 - 2 | 195 | 55 | |
Set Membership: |
|