Assay Details
| Gene Symbol: PRDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714871.3 | XP_002714917.1 | 6 - 7 | 2175 | 60 | |
| XM_008263331.2 | XP_008261553.1 | 6 - 7 | 2190 | 60 | ||
More Information
Set Membership: |
|

PRDM1
Genomic Map