Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273191.2 | XP_008271413.1 | 2 - 3 | 231 | 58 | |
| XM_008273195.2 | XP_008271417.1 | 2 - 3 | 231 | 58 | ||
| XM_017349850.1 | XP_017205339.1 | 2 - 3 | 231 | 58 | ||
| XM_017349851.1 | XP_017205340.1 | 2 - 3 | 231 | 58 | ||
| XM_017349852.1 | XP_017205341.1 | 2 - 3 | 231 | 58 | ||
More Information
Set Membership: |
|

RBMX2
Genomic Map