Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716516.3 | XP_002716562.2 | 2 - 3 | 327 | 80 | |
| XM_017346994.1 | XP_017202483.1 | 3 - 4 | 572 | 80 | ||
More Information
Set Membership: |
|

PLAAT1
Genomic Map