Assay Details
| Gene Symbol: LOC100346895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265233.2 | XP_008263455.1 | 3 - 4 | 431 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100346895| Gene Symbol: LOC100346895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265233.2 | XP_008263455.1 | 3 - 4 | 431 | 69 | |
Set Membership: |
|