Assay Details
| Gene Symbol: KRBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343668.1 | XP_017199157.1 | 551 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRBOX1| Gene Symbol: KRBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343668.1 | XP_017199157.1 | 551 | 66 | ||
Set Membership: |
|