Assay Details
| Gene Symbol: LOC100345328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257790.2 | XP_008256012.2 | 19 - 20 | 2376 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100345328| Gene Symbol: LOC100345328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257790.2 | XP_008256012.2 | 19 - 20 | 2376 | 70 | |
Set Membership: |
|