Assay Details
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715864.3 | XP_002715910.1 | 6 - 7 | 545 | 69 | |
| XM_017346087.1 | XP_017201576.1 | 6 - 7 | 560 | 69 | ||
| XM_017346088.1 | XP_017201577.1 | 6 - 7 | 517 | 69 | ||
More Information
Set Membership: |
|

RPAP2
Genomic Map