Assay Details
| Gene Symbol: APOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256667.2 | XP_008254889.1 | 1 - 2 | 161 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008256667.2 transcripts.
Set Membership: |
|

APOF
Genomic Map