Assay Details
| Gene Symbol: LOC100346672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250749.2 | XP_008248971.1 | 544 | 89 | ||
| XM_008250750.2 | XP_008248972.1 | 552 | 89 | |||
More Information
Set Membership: |
|

LOC100346672
Genomic Map