Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261751.2 | XP_008259973.1 | 2 - 3 | 162 | 84 | |
| XM_008261752.2 | XP_008259974.1 | 2 - 3 | 146 | 84 | ||
| XM_008261753.2 | XP_008259975.1 | 2 - 3 | 145 | 84 | ||
| XM_017344322.1 | XP_017199811.1 | 2 - 3 | 162 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008261751.2, XM_008261752.2, XM_008261753.2, XM_017344322.1 transcripts.
Set Membership: |
|

MTERF1
Genomic Map