Assay Details
| Gene Symbol: CXCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260513.2 | XP_008258735.2 | 1 - 2 | 157 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCR6| Gene Symbol: CXCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260513.2 | XP_008258735.2 | 1 - 2 | 157 | 95 | |
Set Membership: |
|