Assay Details
| Gene Symbol: NMRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708211.3 | XP_002708257.1 | 7 - 8 | 612 | 70 | |
| XM_008256534.2 | XP_008254756.1 | 7 - 8 | 606 | 70 | ||
More Information
Set Membership: |
|

NMRK1
Genomic Map