Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713307.3 | XP_002713353.1 | 2 - 3 | 888 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL6IP5| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713307.3 | XP_002713353.1 | 2 - 3 | 888 | 61 | |
Set Membership: |
|