Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339152.1 | XP_017194641.1 | 2 - 3 | 380 | 68 | |
| XM_017339157.1 | XP_017194646.1 | 2 - 3 | 380 | 68 | ||
More Information
Set Membership: |
|

SEPTIN10
Genomic Map