Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712073.3 | XP_002712119.1 | 8 - 9 | 978 | 72 | |
| XM_008258254.2 | XP_008256476.1 | 7 - 8 | 996 | 72 | ||
| XM_008258258.2 | XP_008256480.1 | 7 - 8 | 818 | 72 | ||
| XM_008258259.2 | XP_008256481.1 | 5 - 6 | 705 | 72 | ||
| XM_017342713.1 | XP_017198202.1 | 8 - 9 | 979 | 72 | ||
More Information
Set Membership: |
|

ASB15
Genomic Map