Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712959.3 | XP_002713005.1 | 3 - 4 | 382 | 63 | |
| XM_008259996.2 | XP_008258218.1 | 3 - 4 | 333 | 63 | ||
| XM_008259997.2 | XP_008258219.1 | 3 - 4 | 383 | 63 | ||
More Information
Set Membership: |
|

SLC25A30
Genomic Map