Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272711.2 | XP_008270933.1 | 2 - 3 | 1030 | 75 | |
| XM_008272712.2 | XP_008270934.1 | 2 - 3 | 949 | 75 | ||
More Information
Set Membership: |
|

ASB12
Genomic Map