Assay Details
| Gene Symbol: LOC100351163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262757.2 | XP_008260979.2 | 1 - 2 | 185 | 105 | |
| XM_017345019.1 | XP_017200508.1 | 1 - 2 | 188 | 105 | ||
More Information
Set Membership: |
|

LOC100351163
Genomic Map