Assay Details
| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262198.2 | XP_008260420.1 | 4 - 5 | 467 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFS4| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262198.2 | XP_008260420.1 | 4 - 5 | 467 | 59 | |
Set Membership: |
|