Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261417.2 | XP_008259639.1 | 7 - 8 | 4452 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF532| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261417.2 | XP_008259639.1 | 7 - 8 | 4452 | 57 | |
Set Membership: |
|