Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709639.3 | XP_002709685.1 | 3 - 4 | 605 | 72 | |
| XM_002709640.3 | XP_002709686.1 | 3 - 4 | 604 | 72 | ||
More Information
Set Membership: |
|

SMYD1
Genomic Map