Assay Details
| Gene Symbol: MRPL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717900.3 | XP_002717946.1 | 1 - 2 | 93 | 57 | |
| XM_017348191.1 | XP_017203680.1 | 1 - 2 | 93 | 57 | ||
| XM_017348192.1 | XP_017203681.1 | 1 - 2 | 93 | 57 | ||
More Information
Set Membership: |
|

MRPL52
Genomic Map