Assay Details
| Gene Symbol: ICMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716152.4 | XP_002716198.1 | 1 - 2 | 282 | 71 | |
| XM_017346620.1 | XP_017202109.1 | 1 - 2 | 284 | 71 | ||
More Information
Set Membership: |
|

ICMT
Genomic Map