Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349156.1 | XP_017204645.1 | 5 - 6 | 1922 | 67 | |
| XM_017349158.1 | XP_017204647.1 | 4 - 5 | 1322 | 67 | ||
More Information
Set Membership: |
|

ZNF652
Genomic Map