Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271736.2 | XP_008269958.1 | 6 - 7 | 817 | 59 | |
| XM_008271738.2 | XP_008269960.1 | 6 - 7 | 853 | 59 | ||
| XM_017349293.1 | XP_017204782.1 | 4 - 5 | 622 | 59 | ||
| XM_017349294.1 | XP_017204783.1 | 6 - 7 | 859 | 59 | ||
More Information
Set Membership: |
|

AMZ2
Genomic Map