Assay Details
| Gene Symbol: PRR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271202.2 | XP_008269424.1 | 10 | 3988 | 128 | |
| XM_008271203.2 | XP_008269425.1 | 10 | 3900 | 128 | ||
More Information
Set Membership: |
|

PRR11
Genomic Map