Assay Details
| Gene Symbol: LOC100350735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711435.3 | XP_002711481.1 | 3 - 4 | 363 | 66 | |
| XM_008257088.2 | XP_008255310.1 | 2 - 3 | 381 | 66 | ||
| XM_017342109.1 | XP_017197598.1 | 3 - 4 | 366 | 66 | ||
| XM_017342110.1 | XP_017197599.1 | 3 - 4 | 323 | 66 | ||
More Information
Set Membership: |
|

LOC100350735
Genomic Map