Assay Details
| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716308.3 | XP_002716354.1 | 3 - 4 | 2120 | 97 | |
| XM_017346855.1 | XP_017202344.1 | 3 - 4 | 2173 | 97 | ||
More Information
Set Membership: |
|

GPR149
Genomic Map