Assay Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248593.2 | XP_008246815.1 | 67 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008248593.2 transcripts.
Set Membership: |
|

TUBAL3
Genomic Map