Assay Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248593.2 | XP_008246815.1 | 461 | 88 | ||
| XM_017337714.1 | XP_017193203.1 | 217 | 88 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBAL3| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248593.2 | XP_008246815.1 | 461 | 88 | ||
| XM_017337714.1 | XP_017193203.1 | 217 | 88 | |||
Set Membership: |
|