Assay Details
| Gene Symbol: MOB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723758.3 | XP_002723804.1 | 441 | 58 | ||
| XM_017339850.1 | XP_017195339.1 | 271 | 58 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017339850.1 transcripts.
Set Membership: |
|

MOB2
Genomic Map