Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712653.3 | XP_002712699.1 | 2 - 3 | 458 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MANSC1| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712653.3 | XP_002712699.1 | 2 - 3 | 458 | 93 | |
Set Membership: |
|