Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255784.2 | XP_008254006.1 | 2 - 3 | 1863 | 72 | |
| XM_008255785.2 | XP_008254007.1 | 2 - 3 | 2056 | 72 | ||
More Information
Set Membership: |
|

ERICH5
Genomic Map