Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708643.3 | XP_002708689.1 | 2 - 3 | 762 | 68 | |
| XM_008259584.2 | XP_008257806.1 | 2 - 3 | 510 | 68 | ||
| XM_008259589.2 | XP_008257811.1 | 2 - 3 | 543 | 68 | ||
| XM_008259598.2 | XP_008257820.1 | 3 - 4 | 555 | 68 | ||
| XM_008259604.2 | XP_008257826.1 | 2 - 3 | 502 | 68 | ||
| XM_017342686.1 | XP_017198175.1 | 2 - 3 | 649 | 68 | ||
More Information
Set Membership: |
|

PUS3
Genomic Map