Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721442.3 | XP_002721488.1 | 253 | 61 | ||
| XM_017337527.1 | XP_017193016.1 | 257 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3BP4| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721442.3 | XP_002721488.1 | 253 | 61 | ||
| XM_017337527.1 | XP_017193016.1 | 257 | 61 | |||
Set Membership: |
|