Assay Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724235.3 | XP_002724281.1 | 1372 | 55 | ||
| XM_017340412.1 | XP_017195901.1 | 1336 | 55 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCDH| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724235.3 | XP_002724281.1 | 1372 | 55 | ||
| XM_017340412.1 | XP_017195901.1 | 1336 | 55 | |||
Set Membership: |
|