Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710019.3 | XP_002710065.1 | 2 - 3 | 245 | 78 | |
| XM_008253703.2 | XP_008251925.1 | 2 - 3 | 245 | 78 | ||
More Information
Set Membership: |
|

ANKRD23
Genomic Map